Assay Details
Target Gene Details
Entrez Gene ID: | 320253 |
Gene Name: | membrane-associated ring finger (C3HC4) 3 |
Gene Aliases: |
6330411I15Rik, A530081L18Rik, AI843639, MARCH-III |
Location: |
Chr.18:56761716-56925652 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
March3 | NM_177115.2 | 5 | 1053 | NP_796089.1 |
AK041088.1 | 5 | 1053 | BAC30814.1 | |
BC107232.1 | 5 | 837 | AAI07233.1 | |
BC107233.1 | 5 | 837 | AAI07234.1 |
More Information
Set Membership: |
Intragenic Exonic |