Assay Details
Target Gene Details
Entrez Gene ID: | 16948 |
Gene Name: | lysyl oxidase |
Gene Aliases: |
AI893619, TSC-160, rrg |
Location: |
Chr.18:52516060-52529867 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lox | NM_001286181.1 | 8 | 3012 | NP_001273110.1 |
NM_001286182.1 | 8 | 2969 | NP_001273111.1 | |
NM_010728.3 | 7 | 3389 | NP_034858.2 | |
AK135213.1 | 7 | 2520 | BAE22461.1 | |
AK141995.1 | 7 | 3275 | BAE24907.1 | |
AK159199.1 | 7 | 3192 | ||
AK159241.1 | 7 | 3168 | BAE34923.1 | |
AK159263.1 | 7 | 3229 | BAE34943.1 | |
AK159368.1 | 7 | 3151 | BAE35026.1 | |
AK164634.1 | 8 | 2970 | BAE37854.1 | |
AK166279.1 | 7 | 3097 | BAE38676.1 | |
BC018439.1 | 7 | 3240 | AAH18439.1 | |
D10837.1 | 1 | 438 | ||
M35796.1 | 1 | 563 | ||
M65142.1 | 7 | 3157 | AAA19032.1 | |
M65143.1 | 4 | 2016 | AAA20185.1 |
More Information
Set Membership: |
Intragenic Exonic |