Assay Details
Target Gene Details
Entrez Gene ID: | 16948 |
Gene Name: | lysyl oxidase |
Gene Aliases: |
AI893619, TSC-160, rrg |
Location: |
Chr.18:52516060-52529867 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lox | NM_001286181.1 | NP_001273110.1 | ||
NM_001286182.1 | NP_001273111.1 | |||
NM_010728.3 | 1 | 281 | NP_034858.2 | |
AK141995.1 | 1 | 167 | BAE24907.1 | |
AK159199.1 | 1 | 85 | ||
AK159241.1 | 1 | 60 | BAE34923.1 | |
AK159263.1 | 1 | 121 | BAE34943.1 | |
AK159368.1 | BAE35026.1 | |||
AK159682.1 | BAE35285.1 | |||
AK159796.1 | 1 | 55 | BAE35377.1 | |
AK160096.1 | 1 | 55 | BAE35626.1 | |
AK164634.1 | BAE37854.1 | |||
AK166279.1 | BAE38676.1 | |||
BB840428.1 | ||||
BC018439.1 | 1 | 132 | AAH18439.1 | |
M65142.1 | AAA19032.1 |
More Information
Set Membership: |
Intragenic Exonic |