Assay Details
Target Gene Details
Entrez Gene ID: | 66119 |
Gene Name: | translocase of outer mitochondrial membrane 6 homolog (yeast) |
Gene Aliases: |
1110002E23Rik, AI663970 |
Location: |
Chr.17:47686645-47688386 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tomm6 | NM_001164729.1 | 3 | 732 | NP_001158201.1 |
NM_025365.3 | 3 | 678 | NP_079641.1 | |
AK003291.1 | 3 | 340 | BAB22694.1 | |
AK010470.1 | 3 | 390 | BAB26963.1 | |
AK019199.1 | 3 | 340 | BAB31597.1 | |
BC021590.1 | 2 | 862 | AAH21590.1 | |
BC037589.1 | 3 | 378 | AAH37589.1 | |
CA319049.1 | 3 | 381 | ||
CA321392.1 | 3 | 262 |
More Information
Set Membership: |
Intragenic Exonic |