Assay Details
Target Gene Details
Entrez Gene ID: | 22247 |
Gene Name: | uridine monophosphate synthetase |
Gene Aliases: |
1700095D23Rik, AA408257, AL033308, BB164745 |
Location: |
Chr.16:33954784-33967302 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Umps | NM_009471.2 | 3 | 664 | NP_033497.1 |
AK035821.1 | 3 | 665 | BAC29199.1 | |
AK035997.1 | 2 | 511 | ||
AK090102.1 | 3 | 667 | ||
AK159808.1 | 3 | 665 | BAE35388.1 | |
AK159955.1 | 3 | 665 | BAE35511.1 | |
BC003887.1 | 3 | 661 | AAH03887.1 | |
BC095981.1 | 3 | 650 | AAH95981.1 | |
M29395.1 | AAA39859.1 |
More Information
Set Membership: |
Intragenic Exonic |