Assay Details
Target Gene Details
Entrez Gene ID: | 30943 |
Gene Name: | protease, serine 30 |
Gene Aliases: |
2010015P21Rik, Disp, Disp1, Tmprss8 |
Location: |
Chr.17:23972127-23980572 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss30 | NM_013921.3 | 4 | 487 | NP_038949.2 |
AJ243866.1 | 4 | 450 | CAB56465.1 | |
AK008263.1 | ||||
BC010970.1 | 4 | 480 | AAH10970.1 | |
BC040348.1 | 4 | 487 | AAH40348.1 |
More Information
Set Membership: |
Intragenic Exonic |