Assay Details
Target Gene Details
Entrez Gene ID: | 13511 |
Gene Name: | desmoglein 2 |
Gene Aliases: |
AA408168, D18Ertd293e |
Location: |
Chr.18:20558116-20604526 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dsg2 | NM_007883.2 | 15 | 3054 | NP_031909.2 |
AB072269.1 | 15 | 3082 | BAB86843.1 | |
AK029320.1 | 4 | 1138 | ||
AK076345.1 | 3 | 895 | BAC36305.1 | |
BC034056.1 | 7 | 1844 | AAH34056.1 | |
BC131653.1 | 15 | 3030 | ||
BC131654.1 | 15 | 3030 |
More Information
Set Membership: |
Intragenic Exonic |