Assay Details
Target Gene Details
Entrez Gene ID: | 56771 |
Gene Name: | mediator complex subunit 20 |
Gene Aliases: |
1110011O05Rik, 2410115I17Rik, AU018348, Trfp |
Location: |
Chr.17:47611596-47624418 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med20 | NM_020048.3 | 4 | 1519 | NP_064432.1 |
AJ245617.1 | 4 | 1484 | CAB53514.1 | |
AK003617.1 | 1 | 581 | ||
AK043396.1 | 4 | 1520 | BAC31536.1 | |
AK146345.1 | 4 | 1520 | BAE27098.1 | |
BC060122.1 | 3 | 5329 | AAH60122.1 |
More Information
Set Membership: |
Intragenic Exonic |