Assay Details
Target Gene Details
Entrez Gene ID: | 11974 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit E |
Gene Aliases: |
Atp6k, Atp6v0e1, M9.2 |
Location: |
Chr.17:26676369-26699647 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp6v0e | NM_025272.2 | 4 | 648 | NP_079548.1 |
AK007610.1 | 4 | 627 | BAB25133.1 | |
AK008464.1 | 4 | 628 | ||
AK013229.1 | 4 | 609 | BAB28727.1 | |
AK017760.1 | 4 | 650 | BAB30911.1 | |
AK030335.1 | 4 | 600 | BAC26909.1 | |
AK159669.1 | 4 | 636 | BAE35273.1 | |
AK168015.1 | 4 | 617 | BAE40002.1 | |
BC021529.1 | 1 | 1142 | AAH21529.1 | |
BC024733.1 | 4 | 612 | AAH24733.1 | |
BY707658.1 | 4 | 624 |
More Information
Set Membership: |
Intragenic Exonic |