Assay Details
Target Gene Details
Entrez Gene ID: | 19123 |
Gene Name: | protein C |
Gene Aliases: |
PC |
Location: |
Chr.18:32123126-32139570 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Proc | NM_001042767.2 | 9 | 1116 | NP_001036232.1 |
NM_001042768.2 | 10 | 1196 | NP_001036233.2 | |
NM_008934.3 | 9 | 1128 | NP_032960.3 | |
NR_045173.1 | 9 | 1132 | ||
AF318182.1 | 9 | 1105 | AAK07918.1 | |
AI118438.1 | 1 | 466 | ||
BC013896.1 | 9 | 1133 | AAH13896.1 | |
CF550816.1 | 5 | 692 | ||
D10445.1 | 8 | 1044 | BAA01235.1 |
More Information
Set Membership: |
Intragenic Exonic |