Assay Details
Target Gene Details
Entrez Gene ID: | 22240 |
Gene Name: | dihydropyrimidinase-like 3 |
Gene Aliases: |
CRMP-4, DRP-3, TUC4, ULIP-1, Ulip, Ulip1 |
Location: |
Chr.18:43320979-43438286 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpysl3 | NM_001291455.1 | NP_001278384.1 | ||
NM_009468.5 | 1 | 157 | NP_033494.1 | |
AF501324.1 | AAQ07444.1 | |||
AK149024.1 | ||||
X87817.1 | 1 | 118 | CAA61082.1 |
More Information
Set Membership: |
Intragenic Exonic |