Assay Details
Target Gene Details
Entrez Gene ID: | 26416 |
Gene Name: | mitogen-activated protein kinase 14 |
Gene Aliases: |
CSBP2, Crk1, Csbp1, Mxi2, PRKM14, PRKM15, p38, p38-alpha, p38MAPK, p38a, p38alpha |
Location: |
Chr.17:28691342-28748405 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mapk14 | NM_001168508.1 | 1 | 444 | NP_001161980.1 |
NM_001168514.1 | 1 | 444 | NP_001161986.1 | |
NM_011951.3 | 1 | 444 | NP_036081.1 | |
AF128892.1 | 1 | 342 | AAF34818.1 | |
AK089059.1 | 1 | 357 | BAC40726.1 | |
AK133684.1 | 1 | 357 | BAE21782.1 | |
BC012235.1 | 1 | 444 | AAH12235.1 | |
BC085303.1 | 1 | 420 | AAH85303.1 | |
D83073.1 | 1 | 275 | BAA19741.1 | |
U10871.1 | 1 | 342 | AAA20888.1 |
More Information
Set Membership: |
Intragenic Exonic |