Assay Details
Target Gene Details
Entrez Gene ID: | 50528 |
Gene Name: | transmembrane protease, serine 2 |
Gene Aliases: |
D16Ertd61e |
Location: |
Chr.16:97564682-97659538 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmprss2 | NM_015775.2 | 14 | 2920 | NP_056590.2 |
AK141350.1 | 14 | 2895 | ||
AK146047.1 | 14 | 2804 | BAE26858.1 | |
BC038393.1 | 14 | 2759 | AAH38393.1 | |
BC054348.1 | 14 | 2920 | AAH54348.1 |
More Information
Set Membership: |
Intragenic Exonic |