Assay Details
Target Gene Details
Entrez Gene ID: | 75599 |
Gene Name: | protocadherin 1 |
Gene Aliases: |
2010005A06Rik, AI585920 |
Location: |
Chr.18:38185914-38212048 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdh1 | NM_029357.3 | 3 | 2014 | NP_083633.2 |
BC024121.1 | 2 | 1565 | AAH24121.1 | |
BC040402.1 | 3 | 2014 | AAH40402.1 |
More Information
Set Membership: |
Intragenic Exonic |