Assay Details
Target Gene Details
Entrez Gene ID: | 77446 |
Gene Name: | HEG homolog 1 (zebrafish) |
Gene Aliases: |
4632417D23Rik, 5530401I02Rik, 9530025L16Rik, BC038614, Gm629, OTTMUSG00000027315 |
Location: |
Chr.16:33684430-33771576 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Heg1 | NM_175256.5 | 2 | 588 | NP_780465.4 |
AK028518.1 | 2 | 589 | BAC25987.1 | |
AK163128.1 | 2 | 589 | BAE37205.1 | |
BB639887.1 | 2 | 589 |