Assay Details
Target Gene Details
Entrez Gene ID: | 70508 |
Gene Name: | bobby sox homolog (Drosophila) |
Gene Aliases: |
5530401J07Rik, 5730403O13Rik |
Location: |
Chr.16:50191844-50432461 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bbx | NM_027444.3 | 17 | 3910 | NP_081720.2 |
AK029747.1 | 17 | 3964 | BAC26596.1 | |
BC066821.1 | 16 | 3627 | AAH66821.1 |
More Information
Set Membership: |
Intragenic Exonic |