Assay Details
Target Gene Details
Entrez Gene ID: | 70508 |
Gene Name: | bobby sox homolog (Drosophila) |
Gene Aliases: |
5530401J07Rik, 5730403O13Rik |
Location: |
Chr.16:50191844-50432461 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bbx | NM_027444.3 | 11 | 1375 | NP_081720.2 |
AF276950.1 | 11 | 1260 | AAL68986.1 | |
AF276951.1 | 11 | 1347 | AAL68987.1 | |
AF276952.1 | 8 | 1103 | AAL68988.1 | |
AF454943.1 | 11 | 1368 | AAL58872.1 | |
AF454944.1 | 9 | 1240 | AAL58873.1 | |
AK029532.1 | 9 | 1242 | BAC26500.2 | |
AK029747.1 | 12 | 1489 | BAC26596.1 | |
AK049516.1 | 11 | 1361 | BAC33788.1 | |
AK050488.1 | 11 | 1314 | BAC34285.1 | |
AK157813.1 | 11 | 1354 | BAE34207.1 | |
AK167724.1 | 11 | 1506 | ||
BC019456.1 | 11 | 1450 | ||
BC024449.1 | 9 | 1223 | ||
BC057869.1 | 11 | 1343 | AAH57869.1 | |
BC066821.1 | 9 | 1002 | AAH66821.1 |
More Information
Set Membership: |
Intragenic Exonic |