Assay Details
Target Gene Details
Entrez Gene ID: | 12013 |
Gene Name: | BTB and CNC homology 1 |
Gene Aliases: |
6230421P05Rik, AI323795 |
Location: |
Chr.16:87698954-87733346 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bach1 | NM_007520.2 | 2 | 243 | NP_031546.1 |
AK030871.1 | 2 | 193 | ||
AK140765.1 | 2 | 244 | BAE24472.1 | |
AK140903.1 | 2 | 244 | BAE24513.1 | |
AK141405.1 | 2 | 178 | BAE24674.1 | |
BC057894.1 | 2 | 164 | AAH57894.1 | |
D86603.1 | 2 | 253 | BAA13137.1 |