Assay Details
Target Gene Details
Entrez Gene ID: | 13196 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain1 |
Gene Aliases: |
AV239055, DEF-1, Ddef1, PAP, mKIAA1249, s19 |
Location: |
Chr.15:64086840-64382919 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asap1 | NM_001276461.1 | 27 | 2881 | NP_001263390.1 |
NM_001276462.1 | 26 | 2961 | NP_001263391.1 | |
NM_001276463.1 | 26 | 2925 | NP_001263392.1 | |
NM_001276467.1 | 28 | 2890 | NP_001263396.1 | |
NM_010026.3 | 29 | 3061 | NP_034156.2 | |
AF075461.1 | 29 | 2880 | AAC98349.1 | |
AF075462.1 | 28 | 2709 | AAC98350.1 | |
AK122477.1 | 27 | 2629 | BAC65759.1 | |
AK146545.1 | 27 | 2719 | BAE27250.1 | |
AK147689.1 | 26 | 2961 | ||
BC002201.1 | 6 | 681 | AAH02201.1 | |
BC048818.1 | 26 | 2891 | AAH48818.1 | |
BC094581.1 | 29 | 3061 | AAH94581.1 | |
BF470337.1 | 4 | 261 | ||
BU707744.1 | 5 | 498 | ||
CO430667.1 | 5 | 460 | ||
U92478.1 | 8 | 873 | AAB82338.1 |