Assay Details
Target Gene Details
Entrez Gene ID: | 668253 |
Gene Name: | deleted in lymphocytic leukemia, 2 |
Gene Aliases: |
1810047A16Rik, 4932411A06Rik, 7730401J12Rik, AI197277, AI788824, Alt1, Gm9069, Leu2 |
Location: |
Chr.14:61602836-61682373 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dleu2 | NR_028264.1 | |||
AF380423.1 | ||||
AF380425.1 | ||||
AK080165.1 | ||||
AK084358.1 | ||||
AK134888.1 | ||||
AK165901.1 |
Target Gene Details
Entrez Gene ID: | 328424 |
Gene Name: | potassium channel regulator |
Gene Aliases: |
Clld4, E030012H22Rik, Gm745 |
Location: |
Chr.14:61607457-61612833 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnrg | NM_001039105.3 | 1 | 433 | NP_001034194.1 |
NM_206974.1 | 1 | 433 | NP_996857.1 | |
AK053138.1 | 1 | 433 | BAC35277.1 | |
AY491736.1 | 1 | 408 | AAS72551.1 | |
AY491737.1 | 1 | 408 | AAS72552.1 | |
BC147091.1 | 1 | 401 | ||
BC147092.1 | 1 | 401 | ||
BC171997.1 | 1 | 401 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |