Assay Details
Target Gene Details
Entrez Gene ID: | 11964 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit A |
Gene Aliases: |
AI647066, Atp6a1, Atp6a2, Atp6v1a1, VA68, VPP2 |
Location: |
Chr.16:44085402-44139700 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp6v1a | NM_007508.5 | 15 | 3245 | NP_031534.2 |
AK150742.1 | 15 | 3230 | ||
AK154869.1 | 15 | 3246 | BAE32890.1 | |
AK160792.1 | 15 | 3260 | BAE36015.1 |
More Information
Set Membership: |
Intragenic Exonic |