Assay Details
Target Gene Details
Entrez Gene ID: | 22045 |
Gene Name: | thyrotropin releasing hormone receptor |
Gene Aliases: |
TRH-R1 |
Location: |
Chr.15:44195924-44235912 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trhr | NM_013696.2 | 4 | 2342 | NP_038724.1 |
BC109005.1 | 4 | 2227 | AAI09006.1 | |
BC109006.1 | 4 | 2227 | AAI09007.1 | |
BC128269.1 | 4 | 2227 | ||
M94384.1 | 5 | 1867 | AAA40437.1 |
More Information
Set Membership: |
Intragenic Exonic |