Assay Details
Target Gene Details
Entrez Gene ID: | 77980 |
Gene Name: | SET binding factor 1 |
Gene Aliases: |
2610510A08Rik, B230113C15Rik, Mtmr5, mKIAA3020 |
Location: |
Chr.15:89288236-89315311 on Build GRCm38 |
Assay Gene Location: | Within Exon 41 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sbf1 | NM_001081030.2 | 40 | 6024 | NP_001074499.2 |
NM_001170561.1 | 41 | 6102 | NP_001164032.1 | |
AK020972.1 | ||||
AK076039.1 | 8 | 1269 | BAC36139.1 | |
AK129485.1 | 40 | 5968 | BAC98295.1 | |
AK147332.1 | 40 | 5998 | ||
BC029156.1 | 11 | 1660 | AAH29156.1 | |
BC098209.1 | 21 | 3367 | AAH98209.1 | |
BC157935.1 | 40 | 5914 | ||
BC158013.1 | 41 | 5992 | ||
BC172094.1 | 40 | 5914 |
More Information
Set Membership: |
Intragenic Exonic |