Assay Details
Target Gene Details
Entrez Gene ID: | 59007 |
Gene Name: | N-glycanase 1 |
Gene Aliases: |
1110002C09Rik, PNGase, Png1 |
Location: |
Chr.14:16249314-16311926 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ngly1 | NM_021504.3 | 12 | 2713 | NP_067479.2 |
AK028248.1 | 12 | 2714 | BAC25839.1 |
More Information
Set Membership: |
Intragenic Exonic |