Assay Details
Target Gene Details
Entrez Gene ID: | 30945 |
Gene Name: | ring finger protein 19A |
Gene Aliases: |
AA032313, Dorfin, Rnf19, UIP117, Ubce7ip2, XYbp |
Location: |
Chr.15:36239933-36283723 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnf19a | NM_013923.2 | 2 | 364 | NP_038951.1 |
AF120207.1 | 1 | 251 | AAF18303.1 | |
AK082665.1 | 2 | 316 | ||
AK147768.1 | 2 | 352 | BAE28125.1 | |
AK166197.1 | 2 | 352 | BAE38624.1 | |
BC040769.1 | 1 | 139 | AAH40769.1 | |
BY207861.1 | 2 | 365 |
More Information
Set Membership: |
Intragenic Exonic |