Assay Details
Target Gene Details
Entrez Gene ID: | 67747 |
Gene Name: | RIB43A domain with coiled-coils 2 |
Gene Aliases: |
4930579A10Rik, Trib |
Location: |
Chr.15:85132060-85144569 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ribc2 | NM_026357.2 | 1 | 94 | NP_080633.2 |
AK006801.1 | 1 | 91 | ||
AK016311.1 | 1 | 95 | BAB30190.2 | |
AK017028.1 | 1 | 115 | ||
BC112373.1 | AAI12374.1 | |||
BY716712.1 | 1 | 115 |
More Information
Set Membership: |
Intragenic Exonic |