Assay Details
Target Gene Details
Entrez Gene ID: | 14573 |
Gene Name: | glial cell line derived neurotrophic factor |
Gene Aliases: |
AI385739 |
Location: |
Chr.15:7810048-7837580 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gdnf | NM_001301332.1 | 3 | 1698 | NP_001288261.1 |
NM_001301333.1 | 3 | 1060 | NP_001288262.1 | |
NM_001301357.1 | 2 | 748 | NP_001288286.1 | |
NM_010275.3 | 3 | 1776 | NP_034405.1 | |
BC119031.1 | 3 | 792 | ||
U37459.1 | 3 | 813 | AAB18672.1 |
More Information
Set Membership: |
Intragenic Exonic |