Assay Details
Target Gene Details
Entrez Gene ID: | 239337 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 12 |
Gene Aliases: |
ADAMTS-12, AI605170 |
Location: |
Chr.15:11064790-11346867 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adamts12 | NM_175501.2 | 2 | 432 | NP_780710.2 |
AJ537452.1 | 2 | 191 | CAD60967.1 | |
AK048612.1 | 2 | 395 | BAC33391.1 | |
AK054015.1 | 2 | 404 | BAC35621.1 | |
AK135354.1 | 2 | 434 | BAE22500.1 |
More Information
Set Membership: |
Intragenic Exonic |