Assay Details
Target Gene Details
Entrez Gene ID: | 20788 |
Gene Name: | sterol regulatory element binding factor 2 |
Gene Aliases: |
AI608257, SREBP-2, SREBP2, SREBP2gc, bHLHd2, lop13, nuc |
Location: |
Chr.15:82147269-82205379 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srebf2 | NM_033218.1 | 19 | 3937 | NP_150087.1 |
AF374267.1 | 19 | 3790 | AAK54763.1 | |
AK141548.1 | 1 | 457 | ||
AK155857.1 | 21 | 3695 | BAE33463.1 | |
AK172219.1 | 19 | 3946 | ||
AK195111.1 | ||||
AK196288.1 | ||||
AK203099.1 | ||||
BG800576.1 |