Assay Details
Target Gene Details
Entrez Gene ID: | 21816 |
Gene Name: | transglutaminase 1, K polypeptide |
Gene Aliases: |
2310004J08Rik, Tgase1 |
Location: |
Chr.14:55700009-55713921 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgm1 | NM_001161714.1 | 12 | 1847 | NP_001155186.1 |
NM_001161715.1 | 12 | 1946 | NP_001155187.1 | |
NM_019984.3 | 12 | 1895 | NP_064368.3 | |
AF186373.1 | 12 | 1838 | AAF35986.1 | |
BC026422.1 | 12 | 1895 | AAH26422.1 |
More Information
Set Membership: |
Intragenic Exonic |