Assay Details
Target Gene Details
Entrez Gene ID: | 28075 |
Gene Name: | desumoylating isopeptidase 1 |
Gene Aliases: |
AI427858, AI850401, D15Wsu75e, DeSI-1, Fam152b, Pppde2 |
Location: |
Chr.15:81992523-82016221 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Desi1 | NM_134095.2 | 6 | 2143 | NP_598856.1 |
AK003172.1 | 1 | 863 | ||
BC022097.1 | 5 | 2098 | AAH22097.1 | |
BC056972.1 | 6 | 2143 | AAH56972.1 | |
BC071205.1 | 6 | 2035 | AAH71205.1 |
Target Gene Details
Entrez Gene ID: | 14375 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 6 |
Gene Aliases: |
70kDa, G22p1, Ku70 |
Location: |
Chr.15:81987826-82040085 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Xrcc6 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |