Assay Details
Target Gene Details
Entrez Gene ID: | 223701 |
Gene Name: | MKL (megakaryoblastic leukemia)/myocardin-like 1 |
Gene Aliases: |
AMKL, Bsac, Mal, Mrtf-A |
Location: |
Chr.15:81012281-81190757 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mkl1 | NM_001082536.1 | 11 | 2134 | NP_001076005.1 |
NM_153049.3 | 10 | 1896 | NP_694629.2 | |
AF385582.1 | 10 | 1667 | AAM94258.1 | |
AF532597.1 | 11 | 2018 | AAN33041.1 | |
AK044188.1 | 11 | 2127 | BAC31809.1 | |
AK089416.1 | 9 | 1726 | BAC40873.1 | |
AK154561.1 | 11 | 2117 | BAE32678.1 | |
AK155821.1 | 11 | 2140 | ||
AK155937.1 | 10 | 1880 | BAE33511.1 | |
BC050941.1 | 11 | 2027 | AAH50941.1 | |
CX562326.1 |
More Information
Set Membership: |
Intragenic Exonic |