Assay Details
Target Gene Details
Entrez Gene ID: | 105653 |
Gene Name: | phytanoyl-CoA hydroxylase interacting protein |
Gene Aliases: |
AW049870, C630010D02Rik, Lnap1ip, PAHX-AP#1, PAHX-AP1 |
Location: |
Chr.14:70457517-70468826 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phyhip | NM_145981.3 | 5 | 2453 | NP_666093.1 |
AK049900.1 | 5 | 2454 | BAC33979.1 | |
BC030494.1 | 5 | 2450 | AAH30494.2 |