Assay Details
Target Gene Details
Entrez Gene ID: | 14390 |
Gene Name: | GA repeat binding protein, alpha |
Gene Aliases: |
GABPalpha |
Location: |
Chr.16:84834925-84863783 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gabpa | NM_008065.2 | 1 | 355 | NP_032091.2 |
AK139916.1 | BAE24181.1 | |||
AK145446.1 | BAE26442.1 | |||
AK145838.1 | 1 | 100 | BAE26687.1 | |
AK148087.1 | 1 | 126 | BAE28337.1 | |
AK150455.1 | BAE29575.1 | |||
AY282793.1 | 1 | 362 | AAQ19478.1 | |
BC052448.1 | 1 | 355 | AAH52448.1 | |
M74515.1 | 1 | 362 | AAA53030.1 |
More Information
Set Membership: |
Intragenic Exonic |