Assay Details
Target Gene Details
Entrez Gene ID: | 20665 |
Gene Name: | SRY (sex determining region Y)-box 10 |
Gene Aliases: |
Dom, Sox21 |
Location: |
Chr.15:79154909-79165235 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sox10 | NM_011437.1 | 3 | 855 | NP_035567.1 |
AF017182.1 | 4 | 917 | AAB99738.1 | |
AF047043.1 | 2 | 621 | AAC24564.1 | |
AF047389.1 | 2 | 621 | AAC24565.1 | |
AK042542.1 | 3 | 856 | ||
AK043220.1 | 3 | 852 | BAC31498.1 | |
BC018551.1 | 2 | 653 | AAH18551.1 | |
BC023356.1 | 3 | 689 | AAH23356.1 | |
BC025171.1 | 3 | 818 | AAH25171.1 | |
BC051058.1 | 3 | 689 | AAH51058.1 | |
U66141.1 | 3 | 852 | AAB49282.1 |