Assay Details
Target Gene Details
Entrez Gene ID: | 223920 |
Gene Name: | sterol O-acyltransferase 2 |
Gene Aliases: |
ACAT2, D15Wsu97e |
Location: |
Chr.15:102150414-102163436 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Soat2 | NM_146064.1 | 6 | 585 | NP_666176.1 |
AF078751.1 | 6 | 555 | AAC64057.1 | |
BC025931.1 | 6 | 585 | AAH25931.1 |
More Information
Set Membership: |
Intragenic Exonic |