Assay Details
Target Gene Details
Entrez Gene ID: | 223722 |
Gene Name: | malonyl CoA:ACP acyltransferase (mitochondrial) |
Gene Aliases: |
AI225907, BC025519 |
Location: |
Chr.15:83546797-83555757 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mcat | NM_001030014.2 | 4 | 1321 | NP_001025185.1 |
AK135457.1 | 4 | 1347 | ||
AK178815.1 | ||||
BC025519.1 | 2 | 817 | AAH25519.1 | |
BC099494.1 | 4 | 1321 | AAH99494.1 |