Assay Details
Target Gene Details
Entrez Gene ID: | 52033 |
Gene Name: | PDZ binding kinase |
Gene Aliases: |
2810434B10Rik, AW538537, D14Ertd732e, TOPK |
Location: |
Chr.14:65805911-65817822 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pbk | NM_023209.2 | 8 | 1327 | NP_075698.1 |
AB041882.1 | 8 | 1330 | BAA99578.1 | |
AK003838.1 | 8 | 1288 | BAB23029.1 | |
AK076121.1 | 8 | 1400 | BAC36199.1 | |
AK217474.1 | 1 | 162 | ||
BC006754.1 | 4 | 692 | AAH06754.1 | |
BC020099.1 | 8 | 1333 | AAH20099.1 |
More Information
Set Membership: |
Intragenic Exonic |