Assay Details
Target Gene Details
Entrez Gene ID: | 67296 |
Gene Name: | suppressor of cytokine signaling 4 |
Gene Aliases: |
3110032M18Rik, A730004F22Rik, AI427843, AU015727, AU015859, Socs7 |
Location: |
Chr.14:47277143-47291591 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Socs4 | NM_080843.2 | 2 | 1106 | NP_543119.2 |
AK014115.1 | 2 | 1137 | BAC25425.1 | |
AY046322.1 | 1 | 934 | AAL03942.1 | |
BC117814.1 | 1 | 1027 | ||
BC117815.1 | 1 | 1027 |
More Information
Set Membership: |
Intragenic Exonic |