Assay Details
Target Gene Details
Entrez Gene ID: | 17174 |
Gene Name: | mannan-binding lectin serine peptidase 1 |
Gene Aliases: |
AW048060, CCPII, Crarf, Masp1/3 |
Location: |
Chr.16:23451785-23521073 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Masp1 | NM_008555.2 | 16 | 2620 | NP_032581.2 |
BC131637.1 | 16 | 2620 | ||
BC131638.1 | 16 | 2620 |
More Information
Set Membership: |
Intragenic Exonic |