Assay Details
Target Gene Details
Entrez Gene ID: | 20512 |
Gene Name: | solute carrier family 1 (glial high affinity glutamate transporter), member 3 |
Gene Aliases: |
AI504299, B430115D02Rik, Eaat1, GLAST, GLAST-1, GLU-T, GluT-1, Gmt1, MGluT1 |
Location: |
Chr.15:8634124-8710807 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc1a3 | NM_148938.3 | 1 | 213 | NP_683740.1 |
BC058711.1 | 1 | 136 | AAH58711.1 | |
BY281420.1 | 1 | 214 |
More Information
Set Membership: |
Intragenic Exonic |