Assay Details
Target Gene Details
Entrez Gene ID: | 54381 |
Gene Name: | carboxypeptidase Q |
Gene Aliases: |
1190003P12Rik, 2610034C17Rik, Hls2, Lal-1, Pgcp |
Location: |
Chr.15:33083129-33594552 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cpq | NM_018755.2 | 3 | 510 | NP_061225.2 |
NM_176073.4 | 2 | 420 | NP_788262.1 | |
AF009513.1 | 2 | 301 | AAC17945.1 | |
AF107835.1 | 2 | 388 | AAD43215.1 | |
AK032972.1 | 2 | 285 | BAC28105.1 | |
AK054422.1 | ||||
AK075686.1 | 2 | 351 | BAC35891.1 | |
AK135096.1 | 2 | 407 | BAE22419.1 | |
AK137164.1 | 2 | 294 | BAE23259.1 | |
AK152139.1 | 3 | 388 | BAE30978.1 | |
AK152145.1 | 3 | 388 | BAE30982.1 | |
AK154361.1 | BAE32537.1 | |||
BC037067.1 | 2 | 267 | AAH37067.1 | |
CK624628.1 | 2 | 420 |
More Information
Set Membership: |
Intragenic Exonic |