Assay Details
Target Gene Details
Entrez Gene ID: | 218756 |
Gene Name: | solute carrier family 4, sodium bicarbonate cotransporter, member 7 |
Gene Aliases: |
E430014N10Rik, NBC3, NBCn1, NBCn1-G |
Location: |
Chr.14:14703011-14799944 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc4a7 | NM_001033270.2 | 1 | 208 | NP_001028442.2 |
AK088400.1 | 1 | 169 | BAC40330.1 | |
AK152233.1 | 1 | 209 | BAE31059.1 | |
AK153567.1 | 1 | 209 | BAE32101.1 | |
GU386352.1 | 1 | 154 | ||
GU386353.2 | 1 | 154 | ||
JQ073566.1 | ||||
JQ349031.1 | 1 | 154 | ||
JQ349032.1 | 1 | 154 | ||
JX254908.1 | 1 | 154 | ||
JX254909.1 | 1 | 154 | ||
JX254910.1 | 1 | 154 | ||
JX254911.1 | 1 | 154 | ||
JX310364.1 | 1 | 154 |
More Information
Set Membership: |
Intragenic Exonic |