Assay Details
Target Gene Details
Entrez Gene ID: | 76872 |
Gene Name: | coiled-coil domain containing 116 |
Gene Aliases: |
4930432J16Rik |
Location: |
Chr.16:17139064-17147146 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc116 | NM_029779.2 | 3 | 373 | NP_084055.2 |
AK015290.1 | 3 | 375 | BAB29781.1 | |
BC050881.1 | 2 | 387 | AAH50881.1 | |
BC057359.1 | 3 | 720 | AAH57359.1 |