Assay Details
Target Gene Details
Entrez Gene ID: | 210126 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma |
Gene Aliases: |
9430020K16Rik, AA959454, AU024130, B130055L10Rik, C79715, D630048H16 |
Location: |
Chr.16:24393350-24992578 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpp | NM_001145952.1 | 7 | 1225 | NP_001139424.1 |
NM_001145954.1 | 3 | 639 | NP_001139426.1 | |
NM_178665.5 | 8 | 1432 | NP_848780.3 | |
AK029335.1 | 8 | 1224 | BAC26401.1 | |
AK029567.1 | 3 | 640 | BAC26516.1 | |
AK040643.1 | 6 | 986 | BAC30654.1 | |
AK045288.1 | 8 | 1293 | BAC32298.1 | |
AK051345.1 | 8 | 1304 | BAC34608.1 | |
AK054550.1 | 7 | 1228 | BAC35821.1 | |
AK076989.1 | 3 | 630 | BAC36552.1 | |
AK085626.1 | 8 | 1424 | ||
AK086307.1 | 8 | 1243 | ||
AK168431.1 | 8 | 1339 | ||
AK169780.1 | 8 | 1244 | BAE41362.1 | |
BC085321.1 | 7 | 1171 | AAH85321.1 |
More Information
Set Membership: |
Intragenic Exonic |