Assay Details
Target Gene Details
Entrez Gene ID: | 75288 |
Gene Name: | solute carrier family 35, member F4 |
Gene Aliases: |
4930550L21Rik |
Location: |
Chr.14:49298519-49525837 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc35f4 | NM_029238.2 | 1 | 226 | NP_083514.1 |
AB041538.1 | 1 | 254 | BAA95023.1 | |
AK034308.1 | 1 | 228 | BAC28670.1 | |
AK043617.1 | 1 | 243 | ||
AK046093.1 | 1 | 436 | BAC32602.1 | |
AK047949.1 | 1 | 223 | BAC33198.1 | |
BC138981.1 | 1 | 125 | ||
BC145720.1 | 1 | 125 |