Assay Details
Target Gene Details
Entrez Gene ID: | 210126 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma |
Gene Aliases: |
9430020K16Rik, AA959454, AU024130, B130055L10Rik, C79715, D630048H16 |
Location: |
Chr.16:24393350-24992578 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpp | NM_178665.5 | 1 | 41 | NP_848780.3 |
CJ166632.1 | 1 | 42 |
Target Gene Details
Entrez Gene ID: | 68841 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma, opposite strand |
Gene Aliases: |
1110054M08Rik, AI481106 |
Location: |
Chr.16:24392556-24393655 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lppos | NR_037954.1 | 1 | 266 | |
AK004252.1 | 1 | 54 |
More Information
Set Membership: |
Intragenic Exonic |