Assay Details
Target Gene Details
Entrez Gene ID: | 97998 |
Gene Name: | DEP domain containing MTOR-interacting protein |
Gene Aliases: |
Depdc6 |
Location: |
Chr.15:55112317-55259273 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Deptor | NM_001037937.3 | 9 | 1356 | NP_001033026.1 |
NM_145470.3 | 9 | 1534 | NP_663445.2 | |
AK139888.1 | 9 | 1356 | BAE24172.1 | |
AK219129.1 | 1 | 198 | ||
AK220300.1 | 9 | 1368 | BAD90225.1 |
More Information
Set Membership: |
Intragenic Exonic |