Assay Details
Target Gene Details
Entrez Gene ID: | 320806 |
Gene Name: | G elongation factor, mitochondrial 2 |
Gene Aliases: |
6530419G12Rik, A930009M04Rik, EF-G2mt, EFG2, MST027, RRF2mt |
Location: |
Chr.13:97137937-97181196 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfm2 | NM_001146043.2 | NP_001139515.1 | ||
NM_001271463.1 | NP_001258392.1 | |||
NM_001271464.1 | NP_001258393.1 | |||
NM_001271465.1 | NP_001258394.1 | |||
NM_177266.5 | NP_796240.3 | |||
AK020100.1 | 1 | 259 | ||
AK028450.1 | BAC25958.1 | |||
AK035819.1 | ||||
AK044371.1 | BAC31889.1 | |||
AK082054.1 | 1 | 264 | ||
AK083441.1 | ||||
AK161797.1 | BAE36579.1 | |||
AK165871.1 | BAE38428.1 | |||
AK167185.1 | ||||
BB841178.1 | ||||
BC027341.1 | AAH27341.1 | |||
BC138233.1 | ||||
BC145208.1 | ||||
BY742160.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |