Assay Details
Target Gene Details
Entrez Gene ID: | 68348 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 1F |
Gene Aliases: |
0610012A11Rik, AI327419, Serpina2, epserin |
Location: |
Chr.12:103688044-103695529 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpina1f | NM_001164742.1 | 5 | 1215 | NP_001158214.1 |
NM_026687.2 | 6 | 1520 | NP_080963.2 | |
AB233454.1 | 6 | 1520 | BAE45638.1 | |
AK002574.1 | 5 | 1408 | BAB22199.1 | |
AK165409.1 | 5 | 1193 | BAE38167.1 | |
BC119526.1 |
More Information
Set Membership: |
Intragenic Exonic |